Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

251232

Sigma-Aldrich

Tetraisopropylthiuram disulfide

98%

Synonym(s):

Bis(diisopropylthiocarbamoyl) disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
([(CH3)2CH]2NC((S)S]2
CAS Number:
Molecular Weight:
352.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

115-117 °C (lit.)

solubility

chloroform: soluble 2.5%, clear, yellow

functional group

amine
disulfide

SMILES string

CC(C)N(C(C)C)C(=S)SSC(=S)N(C(C)C)C(C)C

InChI

1S/C14H28N2S4/c1-9(2)15(10(3)4)13(17)19-20-14(18)16(11(5)6)12(7)8/h9-12H,1-8H3

InChI key

ZUYREEAWHZRZDX-UHFFFAOYSA-N

Application

Tetraisopropylthiuram disulphide has been used:
  • for incorporation of a thiol equivalent into heterocyclic compounds, by reaction with appropriate lithiated heterocycle[1]
  • in preparation of unsymmetrical sulfides[1]
  • as extractant for silver (I) extraction from photographic fixing solution[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New synthesis of unsymmetrical dithia compounds.
Smith K and Tzimas M.
Journal of the Chemical Society. Perkin Transactions 1, 19, 2381-2382 (1995)
Silver extraction for pollution control of photographic fixing solution with tetramethylthiuram disulfide.
Mendoza CS and Kamata S.
Bulletin of the Chemical Society of Japan, 69(12), 3499-3504 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service