Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

250384

Sigma-Aldrich

Ethylpyrazine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
108200
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

refractive index

n20/D 1.498 (lit.)

bp

152-153 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

SMILES string

CCc1cnccn1

InChI

1S/C6H8N2/c1-2-6-5-7-3-4-8-6/h3-5H,2H2,1H3

InChI key

KVFIJIWMDBAGDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethylpyrazine has been used in the synthesis of pyrazinoic acid.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.0 °F

Flash Point(C)

42.78 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notes. Synthesis of Pyrazinoic Acid.
Gainer H.
The Journal of Organic Chemistry, 24(5), 691-691 (1959)
Gustavo Luis Leonardo Scalone et al.
Journal of agricultural and food chemistry, 63(22), 5364-5372 (2015-05-15)
Pyrazines are specific Maillard reaction compounds known to contribute to the unique aroma of many products. Most studies concerning the generation of pyrazines in the Maillard reaction have focused on amino acids, while little information is available on the impact

Protocols

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service