Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

249289

Sigma-Aldrich

Ethyl 2-bromohexanoate

99%

Synonym(s):

Ethyl 2-bromocapronate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CHBrCOOCH2CH3
CAS Number:
Molecular Weight:
223.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.45 (lit.)

bp

213-215 °C (lit.)

density

1.221 g/mL at 25 °C (lit.)

SMILES string

CCCCC(Br)C(=O)OCC

InChI

1S/C8H15BrO2/c1-3-5-6-7(9)8(10)11-4-2/h7H,3-6H2,1-2H3

InChI key

KOUAQOCYMAENKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl 2-bromohexanoate has been used in the preparation of:
  • ethyl 2,2-dibromohexanoate[1]
  • 2-mercaptohexanoic acid derivatives[2]
  • chloroquinoxaline N-oxide[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heiko Zettl et al.
Bioorganic & medicinal chemistry letters, 19(15), 4421-4426 (2009-06-27)
A novel and robust scaffold for highly active PPARalpha agonists based on the 2-mercaptohexanoic acid substructure is presented. Systematic structural variation of the substitution pattern of the phenolic backbone yielded detailed SAR especially of ortho and meta substituents. We corroborated
K S Kim et al.
Journal of medicinal chemistry, 36(16), 2335-2342 (1993-08-06)
A series of novel quinoxaline heterocycle containing angiotensin II receptor antagonist analogs were prepared. This heterocycle was coupled to the biphenyl moiety via an oxygen atom linker instead of a carbon atom. Many of these analogs exhibit very potent activity
Mitsuru Shindo et al.
Chemical & pharmaceutical bulletin, 51(4), 477-478 (2003-04-04)
Aliphatic alpha,alpha-dibromo esters, precursors of ynolates, were synthesized via bromination of lithium alpha-bromo ester enolates with 1,2-dibromotetrafluoroethane in good yields. alpha-Trimethylsilyl-alpha,alpha-dibromo esters were synthesized via radical bromination.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service