Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

249270

1,3-Acetonedicarboxylic acid

98%

Synonym(s):

3-Oxoglutaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2COCH2COOH
CAS Number:
Molecular Weight:
146.10
Beilstein:
1447081
EC Number:
MDL number:

Assay

98%

mp

133 °C (dec.) (lit.)

SMILES string

OC(=O)CC(=O)CC(O)=O

InChI

1S/C5H6O5/c6-3(1-4(7)8)2-5(9)10/h1-2H2,(H,7,8)(H,9,10)

InChI key

OXTNCQMOKLOUAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service