Skip to Content
MilliporeSigma
All Photos(1)

Documents

249238

Sigma-Aldrich

cis-9-Heneicosene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CH=CH(CH2)7CH3
CAS Number:
Molecular Weight:
294.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.45 (lit.)

SMILES string

[H]\C(CCCCCCCC)=C(/[H])CCCCCCCCCCC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service