Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

248177

Sigma-Aldrich

Vitamin E acetate

96%

Synonym(s):

O-Acetyl-α-tocopherol, DL-α-Tocopherol acetate, DL-α-Tocopheryl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H52O3
CAS Number:
Molecular Weight:
472.74
Beilstein:
97512
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

96%

refractive index

n20/D 1.496 (lit.)

bp

224 °C/0.3 mmHg (lit.)

density

0.953 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(OC(C)=O)c(C)c(C)c2O1

InChI

1S/C31H52O3/c1-21(2)13-10-14-22(3)15-11-16-23(4)17-12-19-31(9)20-18-28-26(7)29(33-27(8)32)24(5)25(6)30(28)34-31/h21-23H,10-20H2,1-9H3/t22-,23-,31-/m1/s1

InChI key

ZAKOWWREFLAJOT-CEFNRUSXSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eiko Kato et al.
Bioorganic & medicinal chemistry, 19(21), 6348-6355 (2011-09-29)
The water-soluble vitamin E derivative, sodium dl-α-tocopheryl-6-O-phosphate (1), exhibits protective effects against skin damage. As reported herein, we investigated the actions of 1 on the formation of the inflammatory mediator, prostaglandin E(2) (PGE(2)), as compared to dl-α-tocopheryl acetate (2) and
Dorota Olender et al.
European journal of medicinal chemistry, 44(2), 645-652 (2008-07-02)
Some new nitroimidazole derivatives have been synthesized by treating 4,5-dinitro- and 2-methyl-4,5-dinitroimidazoles with epoxypropane, epichlorohydrin or phenacyl bromide in alkylation reactions. The nitro group in N-substituted 4,5-dinitro- and 2-methyl-4,5-dinitroimidazoles has been replaced with primary and secondary amines to afford 4-amino-5-nitroimidazole
Denis Fourches et al.
Chemical research in toxicology, 23(1), 171-183 (2009-12-18)
Drug-induced liver injury is one of the main causes of drug attrition. The ability to predict the liver effects of drug candidates from their chemical structures is critical to help guide experimental drug discovery projects toward safer medicines. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service