Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

248037

Sigma-Aldrich

2,5-Difluorobenzonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CN
CAS Number:
Molecular Weight:
139.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

33-35 °C (lit.)

SMILES string

Fc1ccc(F)c(c1)C#N

InChI

1S/C7H3F2N/c8-6-1-2-7(9)5(3-6)4-10/h1-3H

InChI key

OJTMHIMQUQOLJV-UHFFFAOYSA-N

General description

Electronic transitions in 2,5-difluorobenzonitrile has been investigated by photoacoustic spectroscopy.

Application

2,5-Difluorobenzonitrile has been used in the preparation of dithiadiazolyl radicals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of molecular packing: crystal structures and magnetic properties of monomeric and dimeric difluorophenyl-1, 2, 3, 5-dithiadiazolyl radicals.
Banister AJ, et al.
J. Chem. Soc., Dalton Trans., 15, 2539-2542 (1997)
Two electronic transitions in the near-UV region of six isomeric difluorobenzonitriles-I.
Ramu K, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 49(2), 223-236 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service