Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

247715

Sigma-Aldrich

4-Amino-N-methylphthalimide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2O2
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

246-248 °C (lit.)

SMILES string

CN1C(=O)c2ccc(N)cc2C1=O

InChI

1S/C9H8N2O2/c1-11-8(12)6-3-2-5(10)4-7(6)9(11)13/h2-4H,10H2,1H3

InChI key

KMEBUNSLFRQSEM-UHFFFAOYSA-N

General description

The intermediate charge-transfer transients of 4-amino-N-methylphthalimide (4AMP) has been characterized by laser flash photolysis, steady-state and time-resolved fluorescence studies. 4AMP is a solvatochromic fluorescent dye. Transient solvation of 4AMP in binary supercritical fluids composed of 2-propanol and CO2 has been studied by frequency-domain fluorescence spectroscopy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of the triplet charge-transfer state of 4-amino-N-methylphthalimide in aprotic and protic media by laser flash photolysis.
Aich S, et al.
J. Chem. Soc., Faraday, 93(17), 2991-2996 (1997)
Reversible excited-state transient solvation in binary supercritical fluids revealed by multifrequency phase and modulation fluorescence.
Betts TA and Bright FV.
Appl. Spectroscopy, 44(7), 1203-1209 (1990)
Axel G Griesbeck et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 2(2), 113-117 (2003-04-01)
The fluorescent 4,5-dimethoxyphthalimides 1-10 were applied as sensors for intra- and intermolecular photoinduced electron transfer processes. Strong intramolecular fluorescence quenching was detected for the thioether 2 and the tertiary amine 3. The fluorescence of the carboxylic acids 4-7 is pH-dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service