Skip to Content
MilliporeSigma
All Photos(1)

Documents

247286

Sigma-Aldrich

Potassium hexahydroxyantimonate

Synonym(s):

Potassium antimonate, Potassium antimonate, hydrated, Potassium pyroantimonate, Potassium pyroantimonate trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KSb(OH)6
CAS Number:
Molecular Weight:
262.90
EC Number:
MDL number:
UNSPSC Code:
12352302

mp

240 °C (dec.) (lit.)

SMILES string

[K+].O[Sb-](O)(O)(O)(O)O

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service