Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

246069

Sigma-Aldrich

2,5-Dichlorobenzotrifluoride

98%

Synonym(s):

2,5-Dichloro-α,α,α-trifluorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CF3
CAS Number:
Molecular Weight:
215.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.483 (lit.)

density

1.483 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

FC(F)(F)c1cc(Cl)ccc1Cl

InChI

1S/C7H3Cl2F3/c8-4-1-2-6(9)5(3-4)7(10,11)12/h1-3H

InChI key

DYBYUWVMLBBEMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Dichlorobenzotrifluoride was used as an internal standard in the quantitative analysis of hexafluoropropylene/vinylidene fluoride copolymers by the CW (Continuous Wave)-NMR method.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of hexafluoropropylene/vinylidene fluoride copolymers by high resolution continuous wave and Fourier transform nuclear magnetic resonance spectrometry.
Brame Jr EG and Yeager FW.
Analytical Chemistry, 48(4), 709-711 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service