Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

245836

Sigma-Aldrich

4,5-Diphenyl-1,3-dioxol-2-one

99%

Synonym(s):

1,2-Diphenylvinylene carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O3
CAS Number:
Molecular Weight:
238.24
Beilstein:
1114120
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

71-75 °C (lit.)

SMILES string

O=C1OC(=C(O1)c2ccccc2)c3ccccc3

InChI

1S/C15H10O3/c16-15-17-13(11-7-3-1-4-8-11)14(18-15)12-9-5-2-6-10-12/h1-10H

InChI key

SROHGOJDCAODGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,5-Diphenyl-1,3-dioxol-2-one may be used in chemical synthesis studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service