Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

244244

Sigma-Aldrich

1-(p-Toluenesulfonyl)imidazole

99%

Synonym(s):

1-Tosylimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2O2S
CAS Number:
Molecular Weight:
222.26
Beilstein:
612451
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

76-78 °C (lit.)

solubility

chloroform: soluble 25 mg/mL, clear, colorless to faintly yellow

SMILES string

Cc1ccc(cc1)S(=O)(=O)n2ccnc2

InChI

1S/C10H10N2O2S/c1-9-2-4-10(5-3-9)15(13,14)12-7-6-11-8-12/h2-8H,1H3

InChI key

YJYMYJRAQYREBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(p-Toluenesulfonyl)imidazole converts alcohols to azides in one step[1].

Application

1-(p-Toluenesulfonyl)imidazole was used in the synthesis of cationic water soluble cyclodextrin, mono-6A-(1-butyl-3-imidazolium)-6A-deoxy-β-cyclodextrin chloride (BIMCD), useful in chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 48, 3445-3445 (2007)
Jae Min Choi et al.
Carbohydrate polymers, 163, 118-128 (2017-03-08)
Ciprofloxacin is a broad-spectrum fluoroquinolone antibiotic used to treat bacterial infections; however, its limited aqueous solubility inhibits its broader clinical uses. This study investigated the complexation effect of mono-6-deoxy-6-aminoethylamino-β-cyclodextrin on the aqueous solubility and bioavailability of ciprofloxacin. During complexation, the
Weihua Tang et al.
Nature protocols, 2(12), 3195-3200 (2007-12-15)
We describe a protocol for the synthesis of mono-6(A)-(1-butyl-3-imidazolium)-6(A)-deoxy-beta-cyclodextrin chloride (BIMCD), a cationic, water-soluble cyclodextrin used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis. Starting from commercially available chemicals, BIMCD is synthesized in five steps.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service