Skip to Content
MilliporeSigma
All Photos(1)

Documents

242675

Sigma-Aldrich

3-Chlorobenzenethiol

97%

Synonym(s):

3-Chlorothiophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SH
CAS Number:
Molecular Weight:
144.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.6021 (lit.)

bp

110 °C/30 mmHg (lit.)

density

1.245 g/mL at 25 °C (lit.)

SMILES string

Sc1cccc(Cl)c1

InChI

1S/C6H5ClS/c7-5-2-1-3-6(8)4-5/h1-4,8H

InChI key

CQJDYPZUDYXHLM-UHFFFAOYSA-N

General description

The transfer constant of 3-chlorobenzenethiol (3-chlorothiophenol) in the polymerization of methyl methacrylate was studied.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of phenols and aromatic thiols on the polymerization of methyl methacrylate.
Canadian Journal of Chemistry, 41(11), 2737-2742 (1963)
Javier Campanini-Salinas et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-22)
A rapid emergence of resistant bacteria is occurring worldwide, endangering the efficacy of antibiotics and reducing the therapeutic arsenal available for treatment of infectious diseases. In the present study, we developed a new class of compounds with antibacterial activity obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service