Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

242004

Sigma-Aldrich

4-Methyl-5-vinylthiazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NS
CAS Number:
Molecular Weight:
125.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.568 (lit.)

bp

78-80 °C/25 mmHg (lit.)

mp

−15 °C (lit.)

density

1.093 g/mL at 25 °C (lit.)

SMILES string

Cc1ncsc1C=C

InChI

1S/C6H7NS/c1-3-6-5(2)7-4-8-6/h3-4H,1H2,2H3

InChI key

QUAMMXIRDIIGDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methyl-5-vinylthiazole is a nitrogen and sulphur-containing heterocyclic compound, present in the species of Annona (magnoliids, Annonaceae) and Caladium bicolor (monocots, Araceae). It is extremely attractive to both male and female beetles of three species of the genus Cyclocephala[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Artur Campos Dália Maia et al.
Journal of chemical ecology, 38(9), 1072-1080 (2012-08-25)
Cyclocephaline scarabs are specialised scent-driven pollinators, implicated with the reproductive success of several Neotropical plant taxa. Night-blooming flowers pollinated by these beetles are thermogenic and release intense fragrances synchronized to pollinator activity. However, data on floral scent composition within such

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service