Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

241822

Sigma-Aldrich

2-Chloro-5-(chloromethyl)thiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4Cl2S
CAS Number:
Molecular Weight:
167.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.575 (lit.)

bp

83-85 °C/8 mmHg (lit.)

density

1.385 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

ClCc1ccc(Cl)s1

InChI

1S/C5H4Cl2S/c6-3-4-1-2-5(7)8-4/h1-2H,3H2

InChI key

MQTKXCOGYOYAMW-UHFFFAOYSA-N

Application

2-Chloro-5-(chloromethyl)thiophene was used in the synthesis of 1-(1,1′-biphenyl-4-yl)-3-(5-chloro-thiophen-2-yl)-2-(1H-imidazol-1-yl)propane and 1-(5-chlorothiophen-2-yl)-2-(1H-imidazol-1-yl)-3-phenylpropane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabrina Castellano et al.
Bioorganic & medicinal chemistry, 16(18), 8349-8358 (2008-09-11)
Several derivatives out of a series of antifungal agents exhibited a good inhibitory potency against aromatase as well as a fairly good selectivity toward CYP17, even if lacking H-bond accepting substituents. Their common structural feature is a flexible backbone that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service