Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

241598

Sigma-Aldrich

2-Methoxyphenyl isocyanate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NCO
CAS Number:
Molecular Weight:
149.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.5414 (lit.)

bp

94.8-95.5 °C/10 mmHg (lit.)

density

1.122 g/mL at 25 °C (lit.)

functional group

isocyanate

SMILES string

COc1ccccc1N=C=O

InChI

1S/C8H7NO2/c1-11-8-5-3-2-4-7(8)9-6-10/h2-5H,1H3

InChI key

SUVCZZADQDCIEQ-UHFFFAOYSA-N

General description

The IR and Raman spectra of 2-methoxyphenyl isocyanate was studied.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jayashree Yenagi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 91, 261-268 (2012-03-03)
IR and Raman spectral measurements in the region 3500-400/50 cm(-1) have been made for the liquid samples of 2-Methoxyphenyl isocyanate and 2-Methoxyphenyl isothiocyanate. A complete assignment of the measured bands has been proposed as aided by conformational and vibration analyses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service