Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

241350

Sigma-Aldrich

Sodium salicylate

ReagentPlus®, ≥99%

Synonym(s):

2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt, Sodium 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COONa
CAS Number:
Molecular Weight:
160.10
Beilstein:
3732792
EC Number:
MDL number:
UNSPSC Code:
12352200

product line

ReagentPlus®

Assay

≥99%

mp

>300 °C (lit.)

SMILES string

[Na+].Oc1ccccc1C([O-])=O

InChI

1S/C7H6O3.Na/c8-6-4-2-1-3-5(6)7(9)10;/h1-4,8H,(H,9,10);/q;+1/p-1

InChI key

ABBQHOQBGMUPJH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

208.9 °F - Pensky-Martens closed cup

Flash Point(C)

98.3 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kui Wang et al.
Journal of medicinal chemistry, 52(20), 6402-6412 (2009-10-16)
Viologens are showing an increasing number of scientific and technical applications in addition to their use as herbicides. However, their high toxicity poses considerable risks to human health, society, and the environment. In this context, we propose a new therapeutic
Sarah L Baldridge et al.
American journal of veterinary research, 72(10), 1305-1317 (2011-10-04)
To determine the pharmacokinetic parameters of xylazine, ketamine, and butorphanol (XKB) administered IM and sodium salicylate (SAL) administered PO to calves and to compare drug effects on biomarkers of pain and distress following sham and actual castration and dehorning. 40
Astam K Patra et al.
Journal of hazardous materials, 201-202, 170-177 (2011-12-16)
We report a highly efficient synthetic strategy for self-assembled mesoporous γ-Al(2)O(3) materials using sodium salicylate as template. The mesoporous γ-Al(2)O(3) samples synthesized following this strategy have high surface areas (231-497 m(2)g(-1)), consist of crystalline tiny spherical nanoparticles of dimensions ca.
Yan-Yan Su et al.
PloS one, 7(10), e46969-e46969 (2012-10-17)
Sodium salicylate (NaSal), an aspirin metabolite, can cause tinnitus in animals and human subjects. To explore neural mechanisms underlying salicylate-induced tinnitus, we examined effects of NaSal on neural activities of the medial geniculate body (MGB), an auditory thalamic nucleus that
Yutaka Sumino et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 85(4 Pt 2), 046307-046307 (2012-06-12)
The dynamics of a viscoelastic Maxwell fluid is studied in a partially filled cylinder rotating around a horizontal axis. At low rotational velocity, the fluid behaves in the same manner as a viscous fluid. A thin fluid film is pulled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service