Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

240710

Sigma-Aldrich

Ethyl chloroacetate

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CO2C2H5
CAS Number:
Molecular Weight:
122.55
Beilstein:
506455
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

10 mmHg ( 38 °C)
3.3 mmHg ( 20 °C)

Assay

≥99%

refractive index

n20/D 1.421 (lit.)

bp

143 °C (lit.)

mp

−26 °C (lit.)

density

1.145 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CCl

InChI

1S/C4H7ClO2/c1-2-7-4(6)3-5/h2-3H2,1H3

InChI key

VEUUMBGHMNQHGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by E16856-ALDRICH | Ethyl chloroacetate 99%

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ethylester of monochloroacetic acid.
C L Braun et al.
Contact dermatitis, 16(2), 114-115 (1987-02-01)
J Q Liu et al.
The Journal of biological chemistry, 272(6), 3363-3368 (1997-02-07)
Asp10 of L-2-haloacid dehalogenase from Pseudomonas sp. YL was proposed to act as a nucleophile to attack the alpha-carbon of L-2-haloalkanoic acids to form an ester intermediate, which is hydrolyzed by nucleophilic attack of a water molecule on the carbonyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service