Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

240702

Sigma-Aldrich

Ethyl acetoacetate

≥99%

Synonym(s):

Acetoacetic ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOC2H5
CAS Number:
Molecular Weight:
130.14
Beilstein:
385838
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

vapor density

4.48 (vs air)

vapor pressure

1 mmHg ( 28.5 °C)

Assay

≥99%

autoignition temp.

580 °F

expl. lim.

9.5 %

bp

181 °C (lit.)

mp

−43 °C (lit.)

density

1.029 g/mL at 20 °C (lit.)

SMILES string

CCOC(=O)CC(C)=O

InChI

1S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3

InChI key

XYIBRDXRRQCHLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

164.3 °F - closed cup

Flash Point(C)

73.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Ting Wang et al.
Journal of biotechnology, 163(3), 292-300 (2012-11-13)
The biocatalytic anti-Prelog stereoselective reduction of ethyl acetoacetate (EAA) to ethyl (R)-3-hydroxybutyrate {(R)-EHB} was successfully conducted in the aqueous system using immobilized Acetobacter sp. CCTCC M209061 cells. Among various microorganisms tested, Acetobacter sp. CCTCC M209061 gave the best performance and
Genomes as resources for biocatalysis.
Jon D Stewart
Advances in applied microbiology, 59, 31-52 (2006-07-11)
Ying-Hui Sun et al.
Journal of biochemical and biophysical methods, 70(6), 850-856 (2008-01-19)
Bacterial beta-ketoacyl-ACP reductase (FabG) and the beta-ketoacyl reductase domain in mammalian fatty acid synthase (FAS) have the same function and both are rendered as the novel targets for drugs. Herein we developed a convenient method, using an available compound ethyl
Gérard Coureaud et al.
Current biology : CB, 16(19), 1956-1961 (2006-10-10)
Mammalian neonates depend on their mother's food supply and use a defined sequence of actions to find her mammary area. Their behavior is initially uncertain and demanding but rapidly becomes optimal. Efficient learning is thus operating in newborns. For instance
Netta Nir et al.
Applied microbiology and biotechnology, 78(4), 659-667 (2008-01-23)
The asymmetric bio-reduction of 4-chloro-acetoacetic-acid-ethyl-ester to the pharmaceutical building block (S)-4-chloro-3-hydroxybutanoate-ethyl-ester requires the utilization of an enantioselective robust biocatalyst. Some of the natural Saccharomyces cerevisiae strains, isolated from Mount Carmel National Park in Israel, were characterized as resistant to environmental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service