Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

240400

Sigma-Aldrich

Dibutyl ether

≥99%

Synonym(s):

Butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3]2O
CAS Number:
Molecular Weight:
130.23
Beilstein:
1732752
MDL number:
Pricing and availability is not currently available.

vapor density

4.48 (vs air)

vapor pressure

4.8 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

365 °F

expl. lim.

8.5 %

refractive index

n20/D 1.399 (lit.)

bp

142-143 °C (lit.)

mp

−98 °C (lit.)

density

0.764 g/mL at 25 °C (lit.)

SMILES string

CCCCOCCCC

InChI

1S/C8H18O/c1-3-5-7-9-8-6-4-2/h3-8H2,1-2H3

InChI key

DURPTKYDGMDSBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service