Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

240028

Sigma-Aldrich

2,6-Di-tert-butyl-4-methylphenol

≥99%

Synonym(s):

2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Beilstein:
1911640
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor density

7.6 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99%

form

solid

autoignition temp.

878 °F

bp

265 °C (lit.)

mp

69-73 °C (lit.)

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Anti-oxidant.

replaced by

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 58, 3597-3597 (1993)
A M Vijesh et al.
European journal of medicinal chemistry, 46(11), 5591-5597 (2011-10-05)
In the present study two new series of Hantzsch 1,4-dihydropyridine derivatives (1,4-DHPs) containing substituted pyrazole moiety (4a-f and 5a-f) were synthesized by the reaction of 3-aryl-1H-pyrazole-4-carbaldehydes with 1,3-dicarbonylcompounds (ethylacetoacetate and methylacetoacetate) and ammonium acetate. The newly synthesized compounds were characterized
Butylated hydroxytoluene (BHT): a review.
H Babich
Environmental research, 29(1), 1-29 (1982-10-01)
Gen-ichiro Arimura et al.
Plant physiology, 146(3), 965-973 (2008-01-01)
Continuous mechanical damage initiates the rhythmic emission of volatiles in lima bean (Phaseolus lunatus) leaves; the emission resembles that induced by herbivore damage. The effect of diurnal versus nocturnal damage on the initiation of plant defense responses was investigated using
Xiao-Juan Duan et al.
Journal of natural products, 70(7), 1210-1213 (2007-07-03)
Four new highly brominated and fully substituted mono- and bis-phenols, 1-(2,3,6-tribromo-4,5-dihydroxybenzyl)pyrrolidin-2-one (1), 1,2-bis(2,3,6-tribromo-4,5-dihydroxyphenyl)ethane (2), 6-(2,3,6-tribromo-4,5-dihydroxybenzyl)-2,5-dibromo-3,4-dihydroxybenzyl methyl ether (3), and 2,3,6-tribromo-4,5-dihydroxybenzyl methyl sulfone (4), were characterized from the marine red alga Symphyocladia latiuscula. In addition, five known bromophenols, bis(2,3,6-tribromo-4,5-dihydroxyphenyl)methane (5), bis(2,3,6-tribromo-4,5-dihydroxybenzyl)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service