Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239917

Butylamine

≥99%

Synonym(s):

1-Aminobutane, n-Butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3NH2
CAS Number:
Molecular Weight:
73.14
Beilstein:
605269
EC Number:
MDL number:

vapor density

2.5 (vs air)

vapor pressure

68 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

594 °F

expl. lim.

9.8 %

refractive index

n20/D 1.401 (lit.)

bp

78 °C (lit.)

mp

−49 °C (lit.)

density

0.74 g/mL at 25 °C (lit.)

SMILES string

CCCCN

InChI

1S/C4H11N/c1-2-3-4-5/h2-5H2,1H3

InChI key

HQABUPZFAYXKJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service