Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239631

Sigma-Aldrich

Acetylsalicylic acid

≥99%

Synonym(s):

2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Beilstein:
779271
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99%

mp

134-136 °C (lit.)

SMILES string

CC(=O)Oc1ccccc1C(O)=O

InChI

1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)

InChI key

BSYNRYMUTXBXSQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks the production of prostaglandins by inhibiting cyclooxygenase (prostaglandin H synthase), with greater selectivity toward the COX-1 isoform. The antithrombotic effect is due to the inhibition of COX-1 in platelets that blocks thromboxane production and platelet aggregation. Chemopreventive against colorectal and other solid tumors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

482.0 °F

Flash Point(C)

250 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Connie N Hess et al.
Journal of the American College of Cardiology, 66(7), 777-787 (2015-08-14)
Bleeding limits anticoagulant treatment in patients with acute coronary syndromes (ACS). We investigated whether background concomitant antiplatelet therapy influences the effects of apixaban after ACS. This study examined high-risk ACS patients who were treated with aspirin or aspirin plus clopidogrel
Niels Hulsman et al.
Journal of medicinal chemistry, 50(10), 2424-2431 (2007-04-20)
Hybrid drug 1 (NO-ASA) continues to attract intense research from chemists and biologists alike. It consists of ASA and a -ONO2 group connected through a spacer and is in preclinical development as an antitumor drug. We report that, contrary to
A O Maree et al.
Journal of thrombosis and haemostasis : JTH, 3(10), 2340-2345 (2005-09-10)
Aspirin (acetylsalicylic acid) irreversibly inhibits platelet cyclooxygenase (COX)-1, the enzyme that converts arachidonic acid (AA) to the potent platelet agonist thromboxane (TX) A2. Despite clear benefit from aspirin in patients with cardiovascular disease (CAD), evidence of heterogeneity in the way
Andrew O Maree et al.
Journal of the American College of Cardiology, 46(7), 1258-1263 (2005-10-04)
We investigated whether use of low-dose enteric-coated (EC) aspirin for secondary prevention of cardiovascular events has sufficient bioavailability to achieve complete platelet cyclooxygenase (COX) inhibition in all individuals. Aspirin reduces cardiovascular morbidity and mortality in patients with pre-existing vascular disease;
Gregg W Stone et al.
Lancet (London, England), 382(9892), 614-623 (2013-07-31)
The relation between platelet reactivity and stent thrombosis, major bleeding, and other adverse events after coronary artery implantation of drug-eluting stents has been incompletely characterised. We aimed to determine the relation between platelet reactivity during dual therapy with aspirin and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service