Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

239534

Sigma-Aldrich

Diisopropylamine

≥99%

Synonym(s):

DIPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHCH(CH3)2
CAS Number:
Molecular Weight:
101.19
Beilstein:
605284
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.5 (vs air)

vapor pressure

50 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

599 °F

expl. lim.

8.5 %

refractive index

n20/D 1.392 (lit.)

bp

84 °C (lit.)

mp

−61 °C (lit.)

density

0.722 g/mL at 25 °C (lit.)

SMILES string

CC(C)NC(C)C

InChI

1S/C6H15N/c1-5(2)7-6(3)4/h5-7H,1-4H3

InChI key

UAOMVDZJSHZZME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

7.8 °F - closed cup

Flash Point(C)

-13.45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koichi Mikami et al.
Chemical record (New York, N.Y.), 6(1), 1-11 (2006-02-14)
The difficulty as well as the significance of the direct generation of metal enolates of alpha-CF(3) ketones cannot be easily understood by chemists unfamiliar with F. In sharp contrast to the sunny side of non-F, hydrocarbon chemistry, F chemistry has
Decoupling deprotonation from metalation: Thia-Fries rearrangement.
Alan M Dyke et al.
Angewandte Chemie (International ed. in English), 47(27), 5067-5070 (2008-05-28)
Songping Liao et al.
Journal of the American Chemical Society, 125(49), 15114-15127 (2003-12-05)
Lithium diisopropylamide-mediated lithiations of N-alkyl ketimines derived from cyclohexanones reveal that simple substitutions on the N-alkyl side chain and the 2-position of the cyclohexyl moiety afford a 60,000-fold range of rates. Detailed rate studies implicate monosolvated monomers at the rate-limiting
Yun Ma et al.
Journal of the American Chemical Society, 132(44), 15610-15623 (2010-10-22)
Lithium diisopropylamide (LDA) in tetrahydrofuran at -78 °C undergoes 1,4-addition to an unsaturated ester via a rate-limiting deaggregation of LDA dimer followed by a post-rate-limiting reaction with the substrate. Muted autocatalysis is traced to a lithium enolate-mediated deaggregation of the
Antonio Ramirez et al.
Journal of the American Chemical Society, 128(31), 10326-10336 (2006-08-03)
Structural, kinetic, and computational studies reveal the mechanistic complexities of a lithium diisopropylamide (LDA)-mediated ester enolization. Hemilabile amino ether MeOCH2CH2NMe2, binding as an eta1 (ether-bound) ligand in the reactant and as an eta2 (chelating) ligand in the transition structure, accelerates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service