Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

238821

Sigma-Aldrich

3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt

99%

Synonym(s):

Sodium 3,5-dichloro-2-hydroxybenzenesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(OH)SO3Na
CAS Number:
Molecular Weight:
265.05
Beilstein:
3640643
EC Number:
MDL number:
UNSPSC Code:
12171500

Assay

99%

SMILES string

[Na+].Oc1c(Cl)cc(Cl)cc1S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Artiss et al.
Clinica chimica acta; international journal of clinical chemistry, 116(3), 301-309 (1981-11-11)
A uricase-peroxidase coupled system, for the determination of uric acid, applied to a CentrifiChem-500 centrifugal fast analyser is described. Relatively large amounts of ascorbic acid, due to the inclusion of an ascorbate oxidase urine diluent, and hemoglobin appear not to
J D Artiss et al.
Clinical biochemistry, 16(6), 334-337 (1983-12-01)
A study into the substitution of sodium 2-hydroxy-3,5-dichlorobenzenesulfonate for phenol in the indicator reaction for an automated glucose procedure is presented. Apart from having physical properties that are more conducive to easy handling than phenol, the former material affords considerably
M W McGowan et al.
Clinical chemistry, 29(8), 1513-1517 (1983-08-01)
A procedure for the enzymatic determination of lecithin and sphingomyelin in aqueous solution is described. The phospholipids are first dissolved in chloroform:methanol (2:1 by vol), the solvent is evaporated, and the residue is redissolved in an aqueous zwitterionic detergent solution.
P Fossati et al.
Clinical chemistry, 29(8), 1494-1496 (1983-08-01)
We describe a new colorimetric method for measuring creatinine in serum and urine. Creatinine hydrolysis is catalyzed by creatinine amidohydrolase, and the creatine so produced is assayed in reactions catalyzed sequentially by creatine amidinohydrolase and sarcosine oxidase in a system
J D Artiss et al.
Clinical chemistry, 30(8), 1389-1392 (1984-08-01)
A colorimetric peroxidase-coupled procedure for determination of creatinine in human serum and urine is described. A 30-s sample pre-treatment with bilirubin oxidase eliminates interference from endogenous bilirubin. The 4-aminoantipyrine-2-hydroxy-3,5-dichlorobenzenesulfonate chromogen system of this method is about fourfold more sensitive than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service