Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

238619

Sigma-Aldrich

3-Methyl-2-butanone

99%

Synonym(s):

Isopropyl methyl ketone, Methyl isopropyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOCH3
CAS Number:
Molecular Weight:
86.13
Beilstein:
1071238
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

form

liquid

refractive index

n20/D 1.388 (lit.)

bp

94-95 °C (lit.)

mp

−92 °C (lit.)

density

0.805 g/mL at 25 °C (lit.)

SMILES string

CC(C)C(C)=O

InChI

1S/C5H10O/c1-4(2)5(3)6/h4H,1-3H3

InChI key

SYBYTAAJFKOIEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F

Flash Point(C)

-1 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huaqing Duan et al.
Organic & biomolecular chemistry, 13(24), 6782-6788 (2015-05-27)
A simple and efficient strategy for the synthesis of indane-1,3-dione derivatives through a palladium(0)-catalyzed reaction incorporating tert-butyl isocyanide has been developed. In addition, by applying this protocol as the key step, indenopyrazole derivatives can be easily synthesized in high yields
Yang Xia et al.
Journal of neuroinflammation, 12, 155-155 (2015-09-05)
Neuroendoscopy is an innovative technique for neurosurgery that can nonetheless result in traumatic brain injury. The accompanying neuroinflammation may lead to secondary tissue damage, which is the major cause of delayed neuronal death after surgery. The present study investigated the
Sami Sajjadifar et al.
Molecules (Basel, Switzerland), 15(4), 2491-2498 (2010-04-30)
Methyl indolenines (4a-c) and(5a-c) were prepared in high yield by a Fischer indole synthesis reaction of o,m-tolylhydrazine hydrochlorides (1a-b) with isopropyl methyl ketone (2) and 2-methylcyclohexanone (3) in acetic acid at room temperature. o,p- Nitrophenylhydrazines (1c-d) were reacted with 2-methylcyclohexanone
E M Gagan et al.
Archives of environmental contamination and toxicology, 52(3), 283-293 (2007-01-27)
Frequently the toxicity of an organic chemical mixture is close to dose-additive, even when the agents are thought to induce toxicity at different molecular sites of action. These findings appear to conflict with the hypothesis that a strictly dose-additive combined
Henry C Tseng et al.
Neurobiology of aging, 36(6), 2201-2212 (2015-03-31)
Primary open-angle glaucoma (POAG) is characterized by progressive neurodegeneration of retinal ganglion cells (RGCs). Why RGCs degenerate in low-pressure POAG remains poorly understood. To gain mechanistic insights, we developed a novel mouse model based on a mutation in human optineurin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service