Skip to Content
MilliporeSigma
All Photos(1)

Documents

238457

Sigma-Aldrich

1-Chlorononane

98%

Synonym(s):

Nonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8Cl
CAS Number:
Molecular Weight:
162.70
Beilstein:
1733625
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.436 (lit.)
n20/D 1.436

bp

202-204 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCl

InChI

1S/C9H19Cl/c1-2-3-4-5-6-7-8-9-10/h2-9H2,1H3

InChI key

RKAMCQVGHFRILV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Chlorononane has been used in the synthesis of 2-bromo[1-14C]1-decanal, affinity labeling probe for the aliphatic aldehyde site of Vibrio harveyi luciferase.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fried et al.
The Journal of biological chemistry, 259(17), 10754-10759 (1984-09-10)
2-Bromo[1-14C]1-decanal was synthesized as an affinity labeling probe for the aliphatic aldehyde site of Vibrio harveyi luciferase. In the presence of excess amounts of this probe, the inactivation of bacterial luciferase occurred following apparent first order kinetics. This inactivation was
Jonas Winters et al.
Physical chemistry chemical physics : PCCP, 21(7), 4053-4062 (2019-02-05)
Polyaramids are a class of high-performance polymers, known for their high mechanical strength and chemical and thermal stability. Their ability to create a network of intermolecular hydrogen bonds causes them to be very poorly soluble in conventional solvents. Hazardous solvents
D Sivaramakrishna et al.
Langmuir : the ACS journal of surfaces and colloids, 31(35), 9546-9556 (2015-08-21)
A homologous series of l-alanine alkyl ester hydrochlorides (AEs) bearing 9-18 C atoms in the alkyl chain have been synthesized and characterized with respect to self-assembly, supramolecular structure, and phase transitions. The CMCs of AEs bearing 11-18 C atoms were
Cuihua Liu et al.
Journal of the science of food and agriculture, 95(1), 111-119 (2014-04-12)
Terpenoids are major components of carotenoids, limonoids and aromas in citrus fruits, resulting in fruit coloration, bitterness and aroma. In this study the carotenoid, limonoid and volatile profiles of red-flesh Chuhong pummelo (CH) and pale green-flesh Feicui pummelo (FC) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service