Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

237752

Sigma-Aldrich

L-Pipecolic acid

Proline homolog., 99%

Synonym(s):

(S)-(−)-2-Piperidinecarboxylic acid, L-Homoproline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
Beilstein:
81093
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

99%

form

powder with small lumps

optical activity

[α]25/D −26.4°, c = 1 in H2O

color

white

mp

272 °C (lit.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H]1CCCCN1

InChI

1S/C6H11NO2/c8-6(9)5-3-1-2-4-7-5/h5,7H,1-4H2,(H,8,9)/t5-/m0/s1

InChI key

HXEACLLIILLPRG-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Occurs in seeds, malt, edible mushrooms, fruits, etc.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Mihalik et al.
Pediatric research, 25(5), 548-552 (1989-05-01)
L-Pipecolic acid, a cyclic imino acid produced during the degradation of lysine, accumulates in body fluids of infants with the generalized peroxisomal disorders, including Zellweger syndrome, neonatal adrenoleukodystrophy, and infantile Refsum disease. Peroxisome-enriched fractions from normal human liver oxidized L-[3H]pipecolic
M R Baumgartner et al.
Annals of neurology, 47(1), 109-113 (2000-01-13)
We describe an 18-year-old patient with psychomotor retardation and abnormally short metatarsi and metacarpals but no other signs of classic Refsum disease. Molecular analysis of the phytanoyl-coenzyme A hydroxylase gene revealed a homozygous deletion causing a frameshift. Surprisingly, L-pipecolic acid
D W Armstrong et al.
Journal of pharmaceutical and biomedical analysis, 11(10), 881-886 (1993-10-01)
Recently it was found that normal adults excrete pipecolic acid primarily as the D-enantiomer even though it is present in the blood stream mainly as the L-enantiomer (i.e. > 98% L). This study of pipecolic acid stereochemistry was extended to
Susan Keay et al.
Chemical biology & drug design, 77(6), 421-430 (2011-03-01)
Interstitial cystitis/painful bladder syndrome is a chronic bladder disorder with epithelial thinning or ulceration, pain, urinary frequency and urgency, for which there is no reliably effective therapy. We previously reported that interstitial cystitis/painful bladder syndrome bladder epithelial cells make a
Kuo-Yuan Hung et al.
The Journal of organic chemistry, 75(24), 8728-8731 (2010-11-26)
An efficient stereoselective synthesis of fully protected (2S,4R)-4-methylpipecolic acid has been developed. The synthesis was achieved by initial asymmetric α-alkylation of glycine with a chiral iodide, affording the linear precursor as a single stereoisomer. Subsequent aldehyde formation using OsO(4)/NaIO(4) followed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service