Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

237345

Sigma-Aldrich

4-Propylaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C6H4NH2
CAS Number:
Molecular Weight:
135.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.543 (lit.)

bp

224-226 °C (lit.)

density

0.919 g/mL at 25 °C (lit.)

SMILES string

CCCc1ccc(N)cc1

InChI

1S/C9H13N/c1-2-3-8-4-6-9(10)7-5-8/h4-7H,2-3,10H2,1H3

InChI key

OAPDPORYXWQVJE-UHFFFAOYSA-N

General description

Oxidation of 4-propylaniline by peroxidase has been studied[1].

Application

4-Propylaniline has been used in the preparation of (E)-2-methyl-4-[(4-propylphenyl)diazenyl]phenol[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies in peroxidase action-XXV: Stereospecificity of peroxidase action.
Baker PB and Saunders BC.
Tetrahedron, 31(16), 1869-1871 (1975)
(E)-2-Methyl-4-[(4-propylphenyl) diazenyl] phenol.
Albayrak C, et al.
Acta Crystallographica Section E, Structure Reports Online, 63(9), o3733-o3733 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service