236918
Chloroform-d
"100%", 99.96 atom % D
Synonym(s):
Deuterochloroform
About This Item
Quality Level
isotopic purity
99.96 atom % D
Assay
≥99.00%
packaging
pk of 10 × 0.5 mL
impurities
≤0.0100% water
water
refractive index
n20/D 1.444 (lit.)
bp
60.9 °C (lit.)
mp
−64 °C (lit.)
density
1.500 g/mL at 25 °C (lit.)
mass shift
M+1
SMILES string
[2H]C(Cl)(Cl)Cl
InChI
1S/CHCl3/c2-1(3)4/h1H/i1D
InChI key
HEDRZPFGACZZDS-MICDWDOJSA-N
Looking for similar products? Visit Product Comparison Guide
accessory
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
Target Organs
Central nervous system, Liver,Kidney
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service