Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

236594

Sigma-Aldrich

1-Methylcyclopropanemethanol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C3H4CH2OH
CAS Number:
Molecular Weight:
86.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.431 (lit.)

bp

128 °C/750 mmHg (lit.)

density

0.887 g/mL at 25 °C (lit.)

SMILES string

CC1(CO)CC1

InChI

1S/C5H10O/c1-5(4-6)2-3-5/h6H,2-4H2,1H3

InChI key

PIZQWRXTMGASCZ-UHFFFAOYSA-N

General description

Kinetics of gas-phase thermal decomposition of methylcyclopropanemethanol has been reported[1].

Application

1-Methylcyclopropanemethanol has been used in the preparation of (±)-2-(benzyloxycarbonylamino)-2-(1′-methylcyclopropyl)ethanamide[2].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evidence for an insertion-homolysis mechanism for carbon-sulphur bond formation in penicillin biosynthesis; 2. Incubation and interpretation.
Baldwin JE, et al.
Tetrahedron, 52(7), 2537-2556 (1996)
Kinetics of the gas-phase thermal decomposition of methylcyclopropanemethanol.
Awan.
J. Chem. Soc. Pak., 27(1), 29-29 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service