Skip to Content
MilliporeSigma
All Photos(2)

Documents

236543

Sigma-Aldrich

N-Methyl-N-(2-pyridyl)formamide

99%

Synonym(s):

N-Formyl-N-methylaminopyridine, Meyers’ Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O
CAS Number:
Molecular Weight:
136.15
Beilstein:
471767
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.566 (lit.)

bp

71-72 °C/0.05 mmHg (lit.)

density

1.137 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)N(C)c1ccccn1

InChI

1S/C7H8N2O/c1-9(6-10)7-4-2-3-5-8-7/h2-6H,1H3

InChI key

DIRVEOCOAHGVJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methyl-N-(2-pyridyl)formamide is also referred to as Meyers formylating agent.

Application

N-Methyl-N-(2-pyridyl)formamide has been used in the synthesis of 3-formyl-2-(4-methoxybenzyl)pyrazol-1-oxide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Halogen dance in pyrazole 1-oxides: synthesis of pyrazolo [3, 4-c] quinoline 1-oxides.
Eskildsen J, et al.
Tetrahedron, 58(38), 7635-7644 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service