Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

236446

Sigma-Aldrich

1-Bromo-3-chloropropane

≥99%

Synonym(s):

I-BCP, Trimethylene bromochloride, Trimethylene chlorobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3Br
CAS Number:
Molecular Weight:
157.44
Beilstein:
605278
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

≥99%

refractive index

n20/D 1.486 (lit.)

bp

144-145 °C (lit.)

density

1.592 g/mL at 25 °C (lit.)

SMILES string

ClCCCBr

InChI

1S/C3H6BrCl/c4-2-1-3-5/h1-3H2

InChI key

MFESCIUQSIBMSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service