Skip to Content
MilliporeSigma
All Photos(1)

Documents

234184

Sigma-Aldrich

Methyl phenylsulfonylacetate

97%

Synonym(s):

Methyl benzenesulfonylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH2COOCH3
CAS Number:
Molecular Weight:
214.24
Beilstein:
2051463
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.535 (lit.)

bp

165 °C/0.05 mmHg (lit.)

density

1.305 g/mL at 20 °C (lit.)

SMILES string

COC(=O)CS(=O)(=O)c1ccccc1

InChI

1S/C9H10O4S/c1-13-9(10)7-14(11,12)8-5-3-2-4-6-8/h2-6H,7H2,1H3

InChI key

NLEAIFBNKPYTGN-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Methyl phenylsulfonylacetate has been used:
  • in a key step during the synthesis of (−)-hybridalactone, marine eicosanoid isolated from the red alga Laurencia hybrida
  • in dehydrative alkylation of alcohols under modified Mitsunobu conditions followed by desulfonylation using magnesium
  • in three step synthesis of 2-(hydroxymethyl)indene

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved synthesis of 2-(hydroxymethyl) indene.
Palandoken H, et al.
Organic Prep. and Proc. Int., 28(6), 702-704 (1996)
Methyl Carboxymethylenation of Alcohols via Dehydrative Alkylation.
YuJ, et al.
Synlett, 1995(11), 1127-1128 (1995)
Koichiro Ota et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(42), 13531-13537 (2012-09-12)
(-)-Hybridalactone (1) is a marine eicosanoid isolated from the red alga Laurencia hybrida. This natural product contains cyclopropane, cyclopentane, 13-membered macrolactone and epoxide ring systems incorporating seven stereogenic centers. Moreover, this compound has an acid-labile skipped Z,Z-diene motif. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service