Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

234060

Sigma-Aldrich

Chromoxane Cyanine R

Dye content 40 %

Synonym(s):

Eriochrome® Cyanine R, Eriochrome® Cyanine RC, Eriochrome® cyanine R, Mordant Blue 3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H15Na3O9S
CAS Number:
Molecular Weight:
536.39
Colour Index Number:
43820
Beilstein:
3873874
EC Number:
MDL number:
UNSPSC Code:
12171500

composition

Dye content, 40%

λmax

512 nm

SMILES string

[Na+].[Na+].[Na+].Cc1cc(cc(C([O-])=O)c1O)\C(=C2\C=C(C)C(=O)C(=C2)C([O-])=O)c3ccccc3S([O-])(=O)=O

Legal Information

Eriochrome is a registered trademark of Huntsman Petrochemical, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service