Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

233854

Sigma-Aldrich

Sodium cacodylate trihydrate

98%

Synonym(s):

Cacodylic acid sodium salt trihydrate, Dimethylarsinic acid sodium salt, Dimethylarsonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2AsO2Na · 3H2O
CAS Number:
Molecular Weight:
214.03
Beilstein:
3702348
EC Number:
MDL number:

Assay

98%

SMILES string

[Na+].[H]O[H].[H]O[H].[H]O[H].C[As](C)([O-])=O

InChI

1S/C2H7AsO2.Na.3H2O/c1-3(2,4)5;;;;/h1-2H3,(H,4,5);;3*1H2/q;+1;;;/p-1

InChI key

RLGWPHBPRCROJO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium cacodylate trihydrate [Na(CH3)2 AsO2 . 3H2O] is a buffering agent typically employed in the preparation of biological samples for electron microscopy.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of diapause and cold acclimation on egg ultrastructure: new insights into the cold hardiness mechanisms of the Asian tiger mosquito Aedes (Stegomyia) albopictus
Kress A, et al.
Journal of Vector Ecology : Journal of the Society for Vector Ecology, 41(1), 142-150 (2016)
Novel buffer systems for macromolecular crystallization
Newman, Janet
Acta Crystallographica. Section F, Structural Biology Communications, 60(3), 610-612 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service