Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

233285

Sigma-Aldrich

3-Chloro-4-fluorophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)OH
CAS Number:
Molecular Weight:
146.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

104 °C/11 mmHg (lit.)

mp

38-40 °C (lit.)

SMILES string

Oc1ccc(F)c(Cl)c1

InChI

1S/C6H4ClFO/c7-5-3-4(9)1-2-6(5)8/h1-3,9H

InChI key

ZQXLIXHVJVAPLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Chloro-4-fluorophenol has been used in the preparation of 3-chloro-4-fluoro-2′-methoxycarbonyldiphenyl ether required for the synthesis of flavone and xanthone derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Mari et al.
Farmaco (Societa chimica italiana : 1989), 54(6), 411-415 (1999-08-12)
A number of flavone and xanthone derivatives bearing some characteristic features of fluoroquinolones such as the fluorine atom and an ortho piperazine ring are described. The new compounds have been tested for possible cytotoxic and antimicrobial activities. Cytotoxicity of both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service