Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

233242

Sigma-Aldrich

5-Fluoro-2-nitrophenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)OH
CAS Number:
Molecular Weight:
157.10
Beilstein:
1870311
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

34-37 °C (lit.)

functional group

fluoro
nitro

SMILES string

Oc1cc(F)ccc1[N+]([O-])=O

InChI

1S/C6H4FNO3/c7-4-1-2-5(8(10)11)6(9)3-4/h1-3,9H

InChI key

QQURWFRNETXFTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Fluoro-2-nitrophenol has been used in:
  • total synthesis of the (+/−)-CC-1065 CPI subunit[1]
  • synthesis of 2-(7-fluoro-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazin-6-yl)-4,5,6,7-tetrahydro-2H-isoindoline-1,3-diones[2]
  • 7-substituted 2H-1,4-benzoxazin-3-(4H)-ones, possessing therapeutic potential as inhibitors of angiogenesis[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael D Ganton et al.
The Journal of organic chemistry, 72(2), 574-582 (2007-01-16)
CC-1065 and the related duocarmycins are members of a structurally unique family of naturally occurring molecules and remain some of the most rigorously studied antitumor compounds to date. Herein we describe a total synthesis of the (+/-)-CC-1065 CPI subunit in
Ming-Zhi Huang et al.
Journal of agricultural and food chemistry, 53(20), 7908-7914 (2005-09-30)
The mode of action of 2-(7-fluoro-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazin-6-yl)-4,5,6,7-tetrahydro-2H-isoindoline-1,3-diones, including the commercial herbicide flumioxazin, had been identified as inhibition of protoporphyrinogen oxidase (protox). As part of continuous efforts to search for new herbicides with high efficacy, broad-spectrum activity, and safety to crops, flumioxazin
Recent advances in the synthesis of 2H-1, 4-benzoxazin-3-(4H)-ones and 3, 4-dihydro-2 H -1, 4-benzoxazines.
Ilas J, et al.
Tetrahedron, 61(31), 7325-7348 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service