Skip to Content
MilliporeSigma
All Photos(1)

Documents

232742

Sigma-Aldrich

3-Hydroxy-6-methyl-2-nitropyridine

99%

Synonym(s):

6-Methyl-2-nitro-3-pyridinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O3
CAS Number:
Molecular Weight:
154.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

106-107 °C (lit.)

SMILES string

Cc1ccc(O)c(n1)[N+]([O-])=O

InChI

1S/C6H6N2O3/c1-4-2-3-5(9)6(7-4)8(10)11/h2-3,9H,1H3

InChI key

WZMGQHIBXUAYGS-UHFFFAOYSA-N

General description

Molecular structure and corresponding vibrational assignments of 3-hydroxy-6-methyl-2-nitropyridine have been investigated using density functional theory (DFT).

Application

3-Hydroxy-6-methyl-2-nitropyridine was used in the synthesis of 3-methoxy-6-methyl-2-nitropyridine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nucleophilic aromatic substitution by [18F] fluoride at substituted 2-nitropyridines.
Malik N, et al.
J. Radioanal. Nucl. Chem., 283(3), 757-764 (2010)
M Karnan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 96, 51-62 (2012-06-05)
The optimized molecular structure and corresponding vibrational assignments of 3-hydroxy-6-methyl-2-nitropyridine have been investigated using density functional theory (DFT) B3LYP method with 6-311++G(d,p), 6-311++G(2d,2p) and 6-311++G(3d,3p) basis sets. Investigation of the relative orientation of the hydroxyl group with respect to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service