Skip to Content
MilliporeSigma
All Photos(2)

Documents

232122

Sigma-Aldrich

2-Aminoperimidine hydrobromide hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9N3 · HBr · xH2O
CAS Number:
Molecular Weight:
264.12 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

297 °C (lit.)

SMILES string

Br[H].[H]O[H].NC1=Nc2cccc3cccc(N1)c23

InChI

1S/C11H9N3.BrH.H2O/c12-11-13-8-5-1-3-7-4-2-6-9(14-11)10(7)8;;/h1-6H,(H3,12,13,14);1H;1H2

InChI key

OOXFTZVZWUMSIW-UHFFFAOYSA-N

Application

2-Aminoperimidine hydrobromide was used as precipitating agent during gravimetric determination of sulphate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfate-selective electrode and its application for sulfate determination in aqueous solutions.
Lomako SV, et al.
Analytica Chimica Acta, 562(2), 216-222 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service