Skip to Content
MilliporeSigma
All Photos(1)

Documents

23030

Sigma-Aldrich

1-Chloroethyl ethyl carbonate

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OCOOCHClCH3
CAS Number:
Molecular Weight:
152.58
Beilstein:
3536477
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

refractive index

n20/D 1.413

bp

159-161 °C (lit.)

density

1.136 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)OC(C)Cl

InChI

1S/C5H9ClO3/c1-3-8-5(7)9-4(2)6/h4H,3H2,1-2H3

InChI key

YVRGKFXJZCTTRB-UHFFFAOYSA-N

Other Notes

Protecting group reagent for introducing the acid-cleavable 1-(ethoxycarbonyloxy)ethyl group in acids, phenols and amines; in pharmaceuticals, the 1-(ethoxycarbonyloxy)ethyl derivatives serve as masked products which are hydrolyzed in plasma

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Svahn et al.
Journal of medicinal chemistry, 29(4), 448-453 (1986-04-01)
Derivatives of the antifibrinolytic drug tranexamic acid [trans-4-(aminomethyl)cyclohexanecarboxylic acid] containing one or two tranexamic acid moieties were synthesized. Most of the derivatives have good stability in acidic and neutral solutions but are easily hydrolyzed in plasma. By measuring the amount
G. Barcelo et al.
Synthesis, 627-627 (1986)
T Nishimura et al.
The Journal of antibiotics, 40(1), 81-90 (1987-01-01)
Orally active 1-(alkyl substituted cyclohexyloxycarbonyloxy)alkyl ester prodrugs (9b-h) of 7 beta-[2-(2-aminothiazol-4-yl)acetamido]-3- [[[1-(2-dimethylaminoethyl)-1H-tetrazol-5-yl]thio]-methyl]ceph+ ++-3- em-4-carboxylic acid (cefotiam, CTM) have been studied as well as the thia (9i) and aza (9j) analogs. These represent derivatives of the 1-(cyclohexylacetoxy)ethyl ester (2) of CTM.
Tigist Y Tamir et al.
Journal of cell science, 133(14) (2020-06-18)
Nuclear factor erythroid 2-related factor 2 (NFE2L2, also known as NRF2) is a transcription factor and master regulator of cellular antioxidant response. Aberrantly high NRF2-dependent transcription is recurrent in human cancer, but conversely NRF2 activity diminishes with age and in
Caiyun G Li et al.
Cell reports, 26(5), 1333-1343 (2019-01-31)
Using proteomic approaches, we uncovered a DNA damage response (DDR) function for peroxisome proliferator activated receptor γ (PPARγ) through its interaction with the DNA damage sensor MRE11-RAD50-NBS1 (MRN) and the E3 ubiquitin ligase UBR5. We show that PPARγ promotes ATM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service