Skip to Content
MilliporeSigma
All Photos(1)

Documents

229660

Sigma-Aldrich

Hafnium(IV) sulfate

≥99.9% trace metals basis (purity excludes 1-2% zirconium)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hf(SO4)2
CAS Number:
Molecular Weight:
370.62
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9% trace metals basis (purity excludes 1-2% zirconium)

form

powder and chunks

reaction suitability

reagent type: catalyst
core: hafnium

impurities

≤1000.0 ppm Trace Metal Analysis

SMILES string

[Hf+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Hf.2H2O4S/c;2*1-5(2,3)4/h;2*(H2,1,2,3,4)/q+4;;/p-4

InChI key

NXKAMHRHVYEHER-UHFFFAOYSA-J

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service