Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

229067

Sigma-Aldrich

Dicinnamalacetone

98%

Synonym(s):

1,9-Diphenyl-1,3,6,8-nonatetraen-5-one

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH=CHCH=CH)2CO
CAS Number:
Molecular Weight:
286.37
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

144-145 °C (lit.)

functional group

ketone
phenyl

SMILES string

O=C(\C=C\C=C\c1ccccc1)/C=C/C=C/c2ccccc2

InChI

1S/C21H18O/c22-21(17-9-7-15-19-11-3-1-4-12-19)18-10-8-16-20-13-5-2-6-14-20/h1-18H/b15-7+,16-8+,17-9+,18-10+

InChI key

RLJALOQFYHCJKG-FVRNMFRHSA-N

Application

Dicinnamalacetone was used as an indicator in measuring the surface acidity of homoionic montmorillonites by titrating certain Hammett indicators adsorbed on the clay with n-butylamine. It was also used as an indicator (yellow to bright red) for detection of excess hydrogen halides in many organic solvents (except alcohols).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface acidity of montmorillonites.
Frenkel M.
Clays and Clay Minerals, 22(5-6), 435-441 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service