Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

227420

Sigma-Aldrich

4-Methyl-5-imidazolemethanol hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N2O · HCl
CAS Number:
Molecular Weight:
148.59
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

97%

mp

233 °C (dec.) (lit.)

SMILES string

Cl.Cc1[nH]cnc1CO

InChI

1S/C5H8N2O.ClH/c1-4-5(2-8)7-3-6-4;/h3,8H,2H2,1H3,(H,6,7);1H

InChI key

UBHDUFNPQJWPRQ-UHFFFAOYSA-N

Application

4-Methyl-5-imidazolemethanol hydrochloride was used in the synthesis of 1,3-bis(5-methyl-4-imidazolyl)-2-thiapropane (tridentate ligand).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

van Steenbergen AC, et al.
J. Chem. Soc., Dalton Trans., 11, 3175-3182 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service