Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

226408

Sigma-Aldrich

o-Methyl-N,N′-diisopropylisourea

97%

Synonym(s):

N,N′-Diisopropyl-O-methylisourea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHC(OCH3)=NCH(CH3)2
CAS Number:
Molecular Weight:
158.24
Beilstein:
1753052
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.436 (lit.)

bp

50-52 °C/0.1 mmHg (lit.)

density

0.871 g/mL at 25 °C (lit.)

functional group

amine

storage temp.

2-8°C

SMILES string

CO\C(NC(C)C)=N/C(C)C

InChI

1S/C8H18N2O/c1-6(2)9-8(11-5)10-7(3)4/h6-7H,1-5H3,(H,9,10)

InChI key

PUVRRPLSJKDMKH-UHFFFAOYSA-N

Application

o-Methyl-N,N′-diisopropylisourea was used in the synthesis of proline betaine (stahydrine) and 4-hydroxyproline betaine (betoncine).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Betaines derived from amino and hydrazino acids as phase transfer catalysts.
Tetrahedron, 46(6), 1911-1922 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service