Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

226386

1-(2-Mesitylenesulfonyl)-1H-1,2,4-triazole

98%

Synonym(s):

MST

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O2S
CAS Number:
Molecular Weight:
251.30
Beilstein:
618223
MDL number:

Assay

98%

SMILES string

Cc1cc(C)c(c(C)c1)S(=O)(=O)n2cncn2

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service