Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

226262

Sigma-Aldrich

4-(Dimethylamino)azobenzene-4′-sulfonyl chloride

96%

Synonym(s):

4-(4-Dimethylaminophenylazo)benzenesulfonyl chloride, DABS-Cl, Dabsyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4N=NC6H4SO2Cl
CAS Number:
Molecular Weight:
323.80
Beilstein:
3064095
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

96%

mp

185 °C (dec.) (lit.)

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccc(cc2)S(Cl)(=O)=O

InChI

1S/C14H14ClN3O2S/c1-18(2)13-7-3-11(4-8-13)16-17-12-5-9-14(10-6-12)21(15,19)20/h3-10H,1-2H3/b17-16+

InChI key

VTVWTPGLLAELLI-WUKNDPDISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chromophoric labeling reagent for amino acids.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Chromatography. B, Biomedical Sciences and Applications, 621, 133-133 (1993)
L Gorbics et al.
Journal of chromatography. A, 676(1), 169-176 (1994-07-29)
The optimal conditions for amino acid analysis of phosphopeptides and N-acetylglucosamine- and N-acetylgalactosamine-containing glycopeptides were investigated by dabsyl-Cl derivatization and reversed-phase high-performance liquid chromatographic separation. By comparing the chromatographic behaviour of the dabsylated phosphoamino acids and dabsylated aminosugars on three
Journal of Chromatography. B, Biomedical Sciences and Applications, 613, 137-137 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service