Skip to Content
MilliporeSigma
All Photos(3)

Documents

225835

Sigma-Aldrich

Resolve-Al YbFOD

Synonym(s):

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium, Yb[fod]3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Yb(OCC(CH3)3CHCOCF2CF2CF3)3
CAS Number:
Molecular Weight:
1061.59
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: erbium
reagent type: catalyst

mp

108-111 °C (lit.)

SMILES string

CC(C)(C)C(\O[Yb](O\C(=C/C(=O)C(F)(F)C(F)(F)C(F)(F)F)C(C)(C)C)O\C(=C/C(=O)C(F)(F)C(F)(F)C(F)(F)F)C(C)(C)C)=C\C(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/3C10H11F7O2.Yb/c3*1-7(2,3)5(18)4-6(19)8(11,12)9(13,14)10(15,16)17;/h3*4,18H,1-3H3;/q;;;+3/p-3/b3*5-4-;

InChI key

KZBQCXBCJMHJOB-VNGPFPIXSA-K

Application

Reactant involved in:
  • Synthesis of rare earth metal complexes for separation of lanthanide metals
  • Conformational mobility of substituted 2-methoxychalcones under the action of lanthanide shift reagents
  • NMR titration on cation-π complexes of bowl-shaped polycyclic aromatic hydrocarbons
  • Specific recognition of chiral amino alcohols

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service