Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

225592

Sigma-Aldrich

2-Methyl-4,5-diphenyloxazole

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13NO
CAS Number:
Molecular Weight:
235.28
MDL number:
UNSPSC Code:
12352100

Assay

95%

refractive index

n20/D 1.628 (lit.)

bp

214 °C/17 mmHg (lit.)

mp

28 °C (lit.)

density

1.116 g/mL at 25 °C (lit.)

SMILES string

Cc1nc(-c2ccccc2)c(o1)-c3ccccc3

Application

Reagent for macrolide synthesis.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 13, 53-53 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service