Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

225320

Sigma-Aldrich

2,3-Dichlorotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3Cl2
CAS Number:
Molecular Weight:
161.03
Beilstein:
2412384
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

refractive index

n20/D 1.551 (lit.)

bp

207-208 °C (lit.)

mp

6 °C (lit.)

density

1.228 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(Cl)c1Cl

InChI

1S/C7H6Cl2/c1-5-3-2-4-6(8)7(5)9/h2-4H,1H3

InChI key

GWLKCPXYBLCEKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riquan Liao et al.
Environmental monitoring and assessment, 191(5), 309-309 (2019-04-28)
To enhance our understanding on environmental conditions of the Maowei Sea in Guangxi province, China, the concentration and distribution of 22 chlorobenzene compounds (CBs) in the surface sediment were determined by gas chromatography-mass spectrometry (GC-MS). The relationship of the sediment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service